missing translation for 'onlineSavingsMsg'
Learn More

Bellingham Stanley Refractometer 60/DR direct reading 230V 50Hz 1.30-1.74 x 0.001

Brand:  Bellingham Stanley 10-99

Refractometer 60/DR direct reading 230V 50Hz 1.30-1.74 x 0.001

Code : 04

Additional Details : Weight : 0.01000kg

Product Code. 12383499

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.