missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Reference™ 2 EU-IVD Single Channel Variable Volume Pipette
Shop All Eppendorf Products
Click to view available options
Volume (Metric):
100 to 1000 μL
2 to 20 μL
Color:
Blue
Gray
Yellow
Unit Size:
Each
Description
Extraordinary precision and accuracy, a long service lifes and an ergonomic design.
Specifications
Specifications
| Certifications/Compliance | In vitro diagnostic device according to Directive 98/79/EC of the European Parliament and the Council dated October 27, 1998 |
| Autoclavable | Autoclavable |
| Color | Yellow |
| Number of Channels | 1 |
| Product Line | Reference™ 2 |
| Sterility | Sterile |
| Type | Pipette |
| Volume (Metric) | 2 to 20 μL |
| Display | 4-digit display visible from all angles |
| For Use With (Application) | Forward pipetting, Reverse pipetting, Removal of supernatants, Sample mixing, Phase extraction, Filling of plates, gels, and reaction vessels |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction