missing translation for 'onlineSavingsMsg'
Learn More

Trajan Scientific and Medical™ Reducing Ferrule for 1/8 in. Fittings

Leak-free system connections for improved chromatography for non-MS systems

Brand:  Trajan Scientific and Medical™ 0726270

Code : 13

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 10045300

  • £40.10 / Pack of 10
Estimated Shipment: 31-05-2024
to see stock.
Add to basket

Description

Description

  • Easy to use
  • Forms a reliable seal
  • Reusable
  • Soft material, forms a soft grip to the capillary column
  • Low emissions
Specifications

Specifications

1/8 in. Fittings
Graphite
Reducing Ferrule
0.8 mm
6.4 mm
6.6 mm
10/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers