missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Color-coded Octahedral PTFE Stir Bar

Use when quick visual identification is highly important. Fisherbrand™ Color-coded Octahedral PTFE Stir Bar is available in three colors: blue, red, and yellow.

Brand:  Fisherbrand™

Code : 41

Additional Details : Weight : 0.02200kg

 View more versions of this product

Product Code. 11888902

  • £6.67 / Pack of 10


Estimated Shipment
10-07-2018
Add to basket

Description

Description

  • Color-coded, octagonal stirrer bars
  • For use where identification is of prime importance
  • Note: Colored PTFE coatings are not as inert as pure PTFE
Specifications

Specifications

Yes
PTFE
PTFE encapsulated Alnico V magnet core
10/Pk.
8mm
Octahedral
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Red
Containers & Vessels / Stirrer Hot Plates
Stir bar
13mm
Videos
Documentation