missing translation for 'onlineSavingsMsg'
Learn More

Hellma™ Glass Standard Cell Cuvettes

Designed to hold samples for spectroscopic experiments and optical analysis. Hellma™ Glass Standard Cell Cuvettes provide optically excellent transparency.

Brand:  Hellma™ 1508736

Code : A0

Additional Details : Weight : 0.03000kg

Product Code. 11746274

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Used in the visible range of the spectrum
  • Low values of refractive index and dispertion
  • Good resistance to chemicals
  • Minimal thermal expansion
  • Manufactured from borosilicate glass
Specifications

Specifications

3.5 mL
2 Pack
45 mm
Glass
360 to 2500 nm
Rectangle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers