missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant SARS SARS Envelope Protein
A cDNA sequence encoding the SARS Envelope was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13417-1mg
Additional Details : Weight : 0.01000kg
Specifications
SARS SARS Envelope Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
50mM Tris-HCl, 60mM NaCl, and 50% glycerol. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
SARS | |
Untagged | |
SARS Envelope protein is >95% pure as determined by 10% PAGE(coomassie staining). |