missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant SARS SARS Envelope Protein

A cDNA sequence encoding the SARS Envelope was constructed and used to recombinantly synthesize the protein.

£173.00 - £1100.00

Specifications

Name SARS SARS Envelope Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity SARS
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15953499
View Documents Promotion Details
enQuireBio™
QP13417-100UG
100 μg
£173.00
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15973499
View Documents Promotion Details
enQuireBio™
QP13417-500UG
500 μg
£660.00
500µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15963499
View Documents Promotion Details
enQuireBio™
QP13417-1MG
1 mg
£1100.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 24-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

SARS SARS Envelope Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
SARS
Untagged
SARS Envelope protein is >95% pure as determined by 10% PAGE(coomassie staining).
Research Use Only
Recombinant Protein
E. coli
50mM Tris-HCl, 60mM NaCl, and 50% glycerol.
Videos
SDS
Documents

Documents

Product Certifications