missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae HYP2 Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) HYP2, was expressed in the hosts and tags indicated.
£281.57 - £2480.00
Specifications
Accession Number | NP_010880.3 |
---|---|
Gene ID (Entrez) | 856677 |
Name | S. cerevisiae HYP2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15969079
|
enQuireBio™
QP5977-EC-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15989079
|
enQuireBio™
QP5977-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15979079
|
enQuireBio™
QP5977-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15999079
|
enQuireBio™
QP5977-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15909089
|
enQuireBio™
QP5977-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15919089
|
enQuireBio™
QP5977-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_010880.3 | |
S. cerevisiae HYP2 Protein | |
Not determined. | |
Full Length | |
S. cerevisiae | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
856677 | |
Research Use Only | |
HYP2 | |
Recombinant Protein | |
E. coli | |
SDEEHTFETA DAGSSATYPM QCSALRKNGF VVIKSRPCKI VDMSTSKTGK HGHAKVHLVA IDIFTGKKLE DLSPSTHNME VPVVKRNEYQ LLDIDDGFLS LMNMDGDTKD DVKAPEGELG DSLQTAFDEG KDLMVTIISA MGEEAAISFK EAARTD | |
Greater than 90% as determined by SDS-PAGE. |