missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus Staphopain B Protein

A DNA sequence encoding the Staphylococcus aureus Staphopain B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9880-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048901

  • £1930.00 / 500µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

WP_001088793.1
500 μg
Not determined.
Full Length
S. aureus
His
Tris-based buffer, 50% glycerol
S. aureus Staphopain B Protein
Research Use Only
sspB
Recombinant Protein
Yeast
DQVQYENTLK NFKIREQQFD NSWCAGFSMA ALLNATKNTD TYNAHDIMRT LYPEVSEQDL PNCSTFPNQM IEYGKSQGRD IHYQEGVPSY EQVDQLTKDN VGIMILAQSV SQNPNDPHLG HALAVVGNAK INDQEKLIYW NPWDTELSIQ DADSSLLHLS FNRDYNWYGS MIGY
Greater than 90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers