missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. aureus Replication initiation Protein
A DNA sequence encoding the Staphylococcus aureus Replication initiation protein, was expressed in the hosts and tags indicated.
£281.57 - £2480.00
Specifications
Accession Number | NP_052693.1 |
---|---|
Gene ID (Entrez) | 3355242 |
Name | S. aureus Replication initiation Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16048370
|
enQuireBio™
QP7349-EC-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16068370
|
enQuireBio™
QP7349-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16058370
|
enQuireBio™
QP7349-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078370
|
enQuireBio™
QP7349-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16088370
|
enQuireBio™
QP7349-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098370
|
enQuireBio™
QP7349-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_052693.1 | |
S. aureus Replication initiation Protein | |
Not determined. | |
Full Length | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
3355242 | |
Research Use Only | |
repD | |
Recombinant Protein | |
E. coli | |
MSTENHSNYL QNKDLDNFSK TGYSNSRLSG NFFTTPQPEL SFDAMTIVGN LNKTNAKKLS DFMSTEPQIR LWDILQTKFK AKALQEKVYI EYDKVKADSW DRRNMRVEFN PNKLTHEEML WLKQNIIDYM EDDGFTRLDL AFDFEDDLSD YYAMTDKAVK KTIFYGRNGK PETKYFGVRD SDRFIRIYNK KQERKDNADV EVMSEHLWRV EIELKRDMVD YWNDCFDDLH ILKPDWTTPE KVKEQAMVYL LLNEEGTWGK LERHAKYKYK QLIKEISPID LTELMKSTLK ENEKQLQKQI DFWQREFRFW K | |
Greater than 90% as determined by SDS-PAGE. |