Learn More
enQuireBio™ Recombinant S. aureus Escherichia coli ssPA / Stringent starvation Protein
Recombinant Protein
Brand:  enQuireBio™ QP5524-5mg
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15966339
  • £2700.00 / 5mg
Estimated Shipment: 03-06-2024
to see stock.
Specifications
WP_000676548.1
5 mg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Recombinant Protein
E. coli
MLPNNDRHQI TDTTNGHYAP VTYIQVEAPT GTFIASGVVV GKDTLLTNKH VVDATHGDPH ALKAFPSAIN QDNYPNGGFT AEQITKYSGE GDLAIVKFSP NEQNKHIGEV VKPATMSNNA ETQVNQNITV TGYPGDKPVA TMWESKGKIT YLKGEAMQYD LSTTGGNSGS PVFNEKNEVI GIHWGGVPNE FNGAVFINEN VRNFLKQNIE DIHFANDDQP NNPDNPDNPN NPDNPNNPDE PNNPDNPNNP DNPDNGDNNN SDNPDAA
Greater than 95% as determined by SDS-PAGE.
S. aureus Escherichia coli ssPA / Stringent starvation Protein
Research Use Only
ssPA
S. aureus
Untagged
10 mM sodium phosphate, pH 7.5