missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rhesus Macaque Rantes Protein
A cDNA sequence encoding the Rantes was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10857-20ug
Additional Details : Weight : 0.01000kg
Specifications
574178 | |
20 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The biological activity determined by a chemotaxis bioassay using human peripheral blood monocytes is in a concentration range of 1.0-10 ng/ml. | |
Rhesus Macaque | |
Untagged | |
Rantes protein was lyophilized from a 0.2microm filtered concentrated solution in 20mM PB pH 6.0 and 500mM NaCl. |
Rantes Protein | |
Research Use Only | |
CCL5 | |
Recombinant Protein | |
E. coli | |
SPHASDTTPC CFAYIARPLP RAHIKEYFYT SGKCSNPAVV FVTRKNRQVC ANPEKKWVRE YINSLEMS | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |