missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rhesus Macaque Rantes Protein

A cDNA sequence encoding the Rantes was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10857-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15955378

  • £2975.00 / 1mg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

574178
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
The biological activity determined by a chemotaxis bioassay using human peripheral blood monocytes is in a concentration range of 1.0-10 ng/ml.
Rhesus Macaque
Untagged
Rantes protein was lyophilized from a 0.2microm filtered concentrated solution in 20mM PB pH 6.0 and 500mM NaCl.
Rantes Protein
Research Use Only
CCL5
Recombinant Protein
E. coli
SPHASDTTPC CFAYIARPLP RAHIKEYFYT SGKCSNPAVV FVTRKNRQVC ANPEKKWVRE YINSLEMS
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers