missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rhesus Macaque IL 3 Protein
A cDNA sequence encoding the IL 3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10711-1mg
Additional Details : Weight : 0.01000kg
Specifications
IL 3 Protein | |
Research Use Only | |
The ED50 as determined by the dose-dependant stimulation of the proliferation of human TF-1 cells is less than 0.1ng/ml, corresponding to a Specific Activity of 10,000,000IU/mg. | |
Rhesus Macaque | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
APMTQTTSLK TSWAKCSNMI DEIITHLNQP PLPSPDFNNL NEEDQTILVE KNLRRSNLEA FSKAVKSLQN ASAIESILKN LPPCLPMATA APTRPPIRIT NGDRNDFRRK LKFYLKTLEN EQAQ | |
Greater than 97.0% as determined by SDS-PAGE. |