missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Tnf alpha Protein
Recombinant Protein
£80.80 - £3085.00
Specifications
Accession Number | NP_036807.1 |
---|---|
Gene ID (Entrez) | 103694380 |
Name | Tnf alpha Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15906289
|
enQuireBio™
QP5508-5UG |
5 μg |
£80.80
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15986279
|
enQuireBio™
QP5508-20UG |
20 μg |
£191.00
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966279
|
enQuireBio™
QP5508-100UG |
100 μg |
£476.00
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15996279
|
enQuireBio™
QP5508-500UG |
500 μg |
£1890.00
500µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15976279
|
enQuireBio™
QP5508-1MG |
1 mg |
£3085.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_036807.1 | |
Tnf alpha Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Cytolysis of mouse L929 cells in the presence of Actinomycin D significantly induced by less than 50 pg/ml corresponding to an activity of at least 2.0 x 10^7 units/mg (or a typical ED50 of less than 10 pg/ml). | |
Rat | |
Untagged | |
10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5 |
103694380 | |
Research Use Only | |
Tnf | |
Recombinant Protein | |
E. coli | |
MLRSSSQNSS DKPVAHVVAN HQAEEQLEWL SQRANALLAN GMDLKDNQLV VPADGLYLIY SQVLFKGQGC PDYVLLTHTV SRFATSYQEK VSLLSAIKSP CPKDTPEGAE LKPWYEPMYL GGVSQLEKGD LLSAEVNLPK YLDITESGQV YFGVIAL | |
Greater than 95% as determined by SDS-PAGE. |