missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Tnf alpha Protein
Recombinant Protein
Brand: enQuireBio™ QP5508-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_036807.1 | |
Tnf alpha Protein | |
Research Use Only | |
Tnf | |
Recombinant Protein | |
E. coli | |
MLRSSSQNSS DKPVAHVVAN HQAEEQLEWL SQRANALLAN GMDLKDNQLV VPADGLYLIY SQVLFKGQGC PDYVLLTHTV SRFATSYQEK VSLLSAIKSP CPKDTPEGAE LKPWYEPMYL GGVSQLEKGD LLSAEVNLPK YLDITESGQV YFGVIAL | |
Greater than 95% as determined by SDS-PAGE. |
103694380 | |
100 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Cytolysis of mouse L929 cells in the presence of Actinomycin D significantly induced by less than 50 pg/ml corresponding to an activity of at least 2.0 x 10^7 units/mg (or a typical ED50 of less than 10 pg/ml). | |
Rat | |
Untagged | |
10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5 |