missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Syndecan-1 / SDC1 / CD138 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Syndecan-1 / SDC1 / CD138, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6656-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_037158.1 | |
Syndecan-1 / SDC1 / CD138 Protein | |
Research Use Only | |
SDC1 | |
Recombinant Protein | |
E. coli | |
QIVTANVPPE DQDGSGDDSD NFSGSGTGAL PDMTLSRQTP STWKDVWLLT ATPTAPEPTS RDTEATLTSI LPAGEKPEEG EPVAHVEAEP DFTARDKEKE ATTRPRETTQ LPVTQQASTA ARATTAQASV TSHPHGDVQP GLHETLAPTA PGQPDHQPPS VEDGGTSVIK EVVEDETTNQ LPAGEGSGEQ DFTFETSGEN TAVAGVEPDL RNQSPVDEGA TGASQGLLDR KEVLG | |
Greater than 90% as determined by SDS-PAGE. |
25216 | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Rat | |
His-GST | |
Tris-based buffer, 50% glycerol |