missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Syndecan-1 / SDC1 / CD138 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Syndecan-1 / SDC1 / CD138, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6656-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16084850

  • £2480.00 / 1mg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_037158.1
Syndecan-1 / SDC1 / CD138 Protein
Research Use Only
SDC1
Recombinant Protein
E. coli
QIVTANVPPE DQDGSGDDSD NFSGSGTGAL PDMTLSRQTP STWKDVWLLT ATPTAPEPTS RDTEATLTSI LPAGEKPEEG EPVAHVEAEP DFTARDKEKE ATTRPRETTQ LPVTQQASTA ARATTAQASV TSHPHGDVQP GLHETLAPTA PGQPDHQPPS VEDGGTSVIK EVVEDETTNQ LPAGEGSGEQ DFTFETSGEN TAVAGVEPDL RNQSPVDEGA TGASQGLLDR KEVLG
Greater than 90% as determined by SDS-PAGE.
25216
1 mg
Not determined.
Extracellular Domain
Rat
His-GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers