missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat SerpinA1 / A1AT Protein

A DNA sequence encoding the Rattus norvegicus (Rat) SerpinA1 / A1AT, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6666-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16004940

  • £545.48 / 100µg

Estimated Shipment
10-11-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Specifications

NP_071964.2
SerpinA1 / A1AT Protein
100 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
24648
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Serpina1
Recombinant Protein
Yeast
EDAQETDTSQ QDQSPTYRKI SSNLADFAFS LYRELVHQSN TSNIFFSPMS ITTAFAMLSL GSKGDTRKQI LEGLEFNLTQ IPEADIHKAF HHLLQTLNRP DSELQLNTGN GLFVNKNLKL VEKFLEEVKN NYHSEAFSVN FADSEEAKKV INDYVEKGTQ GKIVDLMKQL DEDTVFALVN YIFFKGKWKR PFNPEHTRDA DFHVDKSTTV KVPMMNRLGM FDMHYCSTLS SWVLMMDYLG NATAIFLLPD DGKMQHLEQT LTKDLISRFL LNRQTRSAIL YFPKLSISGT YNLKTLLSSL GITRVFNNDA DLSGITEDAP LKLSQAVHKA VLTLDERGTE AAGATVVEAV PMSLPPQVKF DHPFIFMIVE SETQSPLFVG KVIDPTR
Videos
Documents