missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat SDF 1b Protein

A cDNA sequence encoding the SDF 1b was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10863-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15935398

  • £155.00 / 10µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

24772
10 μg
< 1.0 EU per ug protein as determined by the LAL method.
The specific activity as determined by its ability to chemoattract human peripheral blood monocytes at 100-150ng/ml corresponding to a Specific Activity of 6,667-10,000IU/mg.
Rat
Untagged
The Rat SDF1B protein was lyophilized from a concentrated (1 mg/ml) sterile solution containing 20mM Phsophate buffer pH 7.4 and 0.15M NaCl.
SDF 1b Protein
Research Use Only
Cxcl12
Recombinant Protein
E. coli
KPVSLSYRCP CRFFESHVAR ANVKHLKILN TPNCALQIVA RLKSNNRQVC IDPKLKWIQE YLDKALNKRL KM
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers