missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Podocalyxin Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Podocalyxin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9939-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16049051

  • £1406.00 / 500µg

Estimated Shipment: 25-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_620203.1
Podocalyxin Protein
500 μg
Not determined.
Extracellular Domain
Rat
His
Tris-based buffer, 50% glycerol
192181
Greater than 90% as determined by SDS-PAGE.
Research Use Only
PODXL
Recombinant Protein
Yeast
QDNGNKTDTS DITSIDQNQD KPATNQPSNA TPKSSVQPPT PTSISTSSPD PKATQSSNSS VTTTSDSTTD RTSSSTSTVP TTSNSGQTVS SGGKSSDKIT TALPTTLGPV NASSQPTDLN TSTKLPSTPT TNSTASPHQP VSHSEGQHTT VQSSSASVSS SDNTTLLWIL TTSKPTGTSE GTQPIAISTP GITTPVSTPL QPTGSPGGTE SVPTTEEFTH STSSWTPVVS QGPSTPSSTW TSGSYKLKCD PAIKPHEELL ILNLTRDSFC KGSPPNERFL ELLCHSAKAS FKPAEDSCAL ELAPILDNMS -QAVAVKRIV IETKLSPKAV FELLKDKWDD LTEAGVIDIH LGKEGPPEVN EDRFS
Videos
SDS
Documents
Certificates