missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat PKLR / PKRL Protein
A DNA sequence encoding the Rattus norvegicus (Rat) PKLR / PKRL, was expressed in the hosts and tags indicated.
£223.39 - £2480.00
Specifications
Accession Number | NP_036756.3 |
---|---|
Gene ID (Entrez) | 24651 |
Name | PKLR / PKRL Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16003740
|
enQuireBio™
QP6498-EC-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16023740
|
enQuireBio™
QP6498-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16013740
|
enQuireBio™
QP6498-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16033740
|
enQuireBio™
QP6498-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16043740
|
enQuireBio™
QP6498-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16053740
|
enQuireBio™
QP6498-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_036756.3 | |
PKLR / PKRL Protein | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
24651 | |
Research Use Only | |
Pklr | |
Recombinant Protein | |
E. coli | |
MSVQENTLPQ QLWPWIFRSQ KDLAKSALSG APGGPAGYLR RASVAQLTQE LGTAFFQQQQ LPAAMADTFL EHLCLLDIDS QPVAARSTSI IATIGPASRS VDRLKEMIKA GMNIARLNFS HGSHEYHAES IANIREATES FATSPLSYRP VAIALDTKGP EIRTGVLQGG PESEVEIVKG SQVLVTVDPK FQTRGDAKTV WVDYHNITRV VAVGGRIYID DGLISLVVQK IGPEGLVTEV EHGGILGSRK GVNLPNTEVD LPGLSEQDLL DLRFGVQHNV DIIFASFVRK ASDVLAVRDA LGPEGQNIKI ISKIENHEGV KKFDEILEVS DGIMVARGDL GIEIPAEKVF LAQKMMIGRC NLAGKPVVCA TQMLESMITK ARPTRAETSD VANAVLDGAD CIMLSGETAK GSFPVEAVMM QHAIAREAEA AVYHRQLFEE LRRAAPLSRD PTEVTAIGAV EASFKCCAAA IIVLTKTGRS AQLLSQYRPR AAVIAVTRSA QAARQVHLSR GVFPLLYREP PEAIWADDVD RRVQFGIESG KLRGFLRVGD LVIVVTGWRP GSGYTNIMRV LSVS | |
Greater than 90% as determined by SDS-PAGE. |