missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat MMP-2 / CLG4A Protein
A DNA sequence encoding the Rattus norvegicus (Rat) MMP-2 / CLG4A Protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6369-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_112316.2 | |
MMP-2 / CLG4A Protein | |
Research Use Only | |
Mmp2 | |
Recombinant Protein | |
Yeast | |
YNFFPRKPKW DKNQITYRII GYTPDLDPET VDDAFARALK VWSDVTPLRF SRIHDGEADI MINFGRWEHG DGYPFDGKDG LLAHAFAPGT GVGGDSHFDD DELWTLGEGQ VVRVKYGNAD GEYCKFPFLF NGREYSSCTD TGRSDGFLWC STTYNFEKDG KYGFCPHEAL FTMGGNGDGQ PCKFPFRFQG TSYNSCTTEG RTDGYRWCGT TEDYDRDKKY GFCPETAMST VGGNSEGAPC VFPFTFLGNK YESCTSAGRS DGKVWCATTT NYDDDRKWGF CPDQGYSLFL VAAHEFGHAM GLEHSQDPGA LMAPIYTYTK NFRLSNDDIK GIQELYGPSP DADTDTGTGP TPTLGPVTPE ICKQDIVFDG IAQIRGEIFF FKDRFIWRTV TPRDKPTGPL LVATFWPELP EKIDAVYEAP QEEKAVFFAG NEYWVYSAST LERGYPKPLT SLGLPPDVQQ VDAAFNWSKN KKTYIFSGDK FWRYNEVKKK MDPGFPKLIA DSWNAIPDNL DAVVDLQGGG HSYFFKGAYY LKLENQSLKS VKFGSIKSDW LGC | |
Greater than 90% as determined by SDS-PAGE. |
81686 | |
1 mg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |