missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat MMP-2 / CLG4A Protein
A DNA sequence encoding the Rattus norvegicus (Rat) MMP-2 / CLG4A Protein, was expressed in the hosts and tags indicated.
£223.39 - £2410.48
Specifications
Accession Number | NP_112316.2 |
---|---|
Gene ID (Entrez) | 81686 |
Name | MMP-2 / CLG4A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16082800
|
enQuireBio™
QP6369-YE-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16022810
|
enQuireBio™
QP6369-YE-500UG |
500 μg |
£1600.00
500µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16092800
|
enQuireBio™
QP6369-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16012810
|
enQuireBio™
QP6369-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16032810
|
enQuireBio™
QP6369-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16002810
|
enQuireBio™
QP6369-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_112316.2 | |
MMP-2 / CLG4A Protein | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
81686 | |
Research Use Only | |
Mmp2 | |
Recombinant Protein | |
Yeast | |
YNFFPRKPKW DKNQITYRII GYTPDLDPET VDDAFARALK VWSDVTPLRF SRIHDGEADI MINFGRWEHG DGYPFDGKDG LLAHAFAPGT GVGGDSHFDD DELWTLGEGQ VVRVKYGNAD GEYCKFPFLF NGREYSSCTD TGRSDGFLWC STTYNFEKDG KYGFCPHEAL FTMGGNGDGQ PCKFPFRFQG TSYNSCTTEG RTDGYRWCGT TEDYDRDKKY GFCPETAMST VGGNSEGAPC VFPFTFLGNK YESCTSAGRS DGKVWCATTT NYDDDRKWGF CPDQGYSLFL VAAHEFGHAM GLEHSQDPGA LMAPIYTYTK NFRLSNDDIK GIQELYGPSP DADTDTGTGP TPTLGPVTPE ICKQDIVFDG IAQIRGEIFF FKDRFIWRTV TPRDKPTGPL LVATFWPELP EKIDAVYEAP QEEKAVFFAG NEYWVYSAST LERGYPKPLT SLGLPPDVQQ VDAAFNWSKN KKTYIFSGDK FWRYNEVKKK MDPGFPKLIA DSWNAIPDNL DAVVDLQGGG HSYFFKGAYY LKLENQSLKS VKFGSIKSDW LGC | |
Greater than 90% as determined by SDS-PAGE. |