missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat M-CSF / CSF-1 Protein
Recombinant Protein
£80.80 - £5400.00
Specifications
Name | M-CSF / CSF-1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Gene Symbol | CSF1 |
Biological Activity | Rat M-CSF / CSF-1 induces NFS-60 cell proliferation (typical ED50 is ? 2 ng/ml). |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15946329
|
enQuireBio™
QP5518-1UG |
1 μg |
£80.80
1µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15926329
|
enQuireBio™
QP5518-10UG |
10 μg |
£191.00
10µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15916329
|
enQuireBio™
QP5518-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956329
|
enQuireBio™
QP5518-500UG |
500 μg |
£3285.00
500µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15936329
|
enQuireBio™
QP5518-1MG |
1 mg |
£5400.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
M-CSF / CSF-1 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Rat M-CSF / CSF-1 induces NFS-60 cell proliferation (typical ED50 is ? 2 ng/ml). | |
Rat | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |
Research Use Only | |
CSF1 | |
Recombinant Protein | |
E. coli | |
MEVSEHCSHM IGNGHLQILQ QLIDSQMETA CLIEYKFVDQ EQLDDPVCYL KKAFVLVQVI IEETMRFKDN TPNANATERL QELSMKLNSC FIKDYKEQNE ACVQTYKESP LRLLEKIKNF FNETKNFLEK DWNIFSKNCN DSLAKCSSRD VVTKP | |
Greater than 95% as determined by SDS-PAGE. |