missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL6 / Interleukin-6 Protein
Recombinant Protein
£80.80 - £5400.00
Specifications
Accession Number | NP_036721.1 |
---|---|
Gene ID (Entrez) | 24498 |
Name | IL6 / Interleukin-6 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15936269
|
enQuireBio™
QP5501-2UG |
2 μg |
£80.80
2µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15916269
|
enQuireBio™
QP5501-10UG |
10 μg |
£191.00
10µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15906269
|
enQuireBio™
QP5501-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15946269
|
enQuireBio™
QP5501-500UG |
500 μg |
£3285.00
500µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15926269
|
enQuireBio™
QP5501-1MG |
1 mg |
£5400.00
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_036721.1 | |
IL6 / Interleukin-6 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Rat IL6 / Interleukin-6 induces 7TD1 cell proliferation (typical ED50 is ? 10 pg/ml). | |
Rat | |
Untagged | |
0.1% Trifluoroacetic Acid (TFA) + 2:1 mannitol to protein |
24498 | |
Research Use Only | |
IL6 | |
Recombinant Protein | |
E. coli | |
MFPTSQVRRG DFTEDTTHNR PVYTTSQVGG LITYVLREIL EMRKELCNGN SDCMNSDDAL SENNLKLPEI QRNDGCFQTG YNQEICLLKI CSGLLEFRFY LEFVKNNLQD NKKDKARVIQ SNTETLVHIF KQEIKDSYKI VLPTPTSNAL LMEKLESQKE WLRTKTIQLI LKALEEFLKV TMRSTRQT | |
Greater than 90 as determined by SDS-PAGE. |