missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat IL11RA Protein

A cDNA sequence encoding the IL11RA was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12403-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15950309

  • £155.00 / 5µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

IL11RA Protein
Research Use Only
Recombinant Protein
Baculovirus
TPCPQAWGPP GVQYGQPGRP VMLCCPGVNA GTPVSWFRDG DSRLLQGPDS GLGHRLVLAQ VDSRDEGTYV CRTLDGVFGG MVTLKLGSPP ARPEVSCQAV DYENFSCTWS PGRVSGLPTR YLTSYRKKTL PGAESQRESP STGPWPCPQD PLEASRCVVH GAEFWSEYRI NVTEVNPLGA STCLLDVRLQ RILRPDPPQG LRVESVPGYP RRLHASWTYP ASWRRQPHFL LKFRLQYRPA QHPAWSTVEP IGLEELITDA VAGLPHAVRV SARDFLDAGT WSAWSPEAWG TPSTGPLRDE VPDGSRGHEQ KLEAAAQEDS PAPPSPSLQP DPRPLDHRDP LEQVAVLAVE HHHHHH
Greater than 85.0% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Rat
His
IL11RA protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers