missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL11RA Protein
A cDNA sequence encoding the IL11RA was constructed and used to recombinantly synthesize the protein.
£99.15 - £1320.00
Specifications
Name | IL11RA Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Rat |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15930309
|
enQuireBio™
QP12403-1UG |
1 μg |
£99.15
1µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15950309
|
enQuireBio™
QP12403-5UG |
5 μg |
£155.00
5µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15940309
|
enQuireBio™
QP12403-50UG |
50 μg |
£1320.00
50µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
IL11RA Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Rat | |
His | |
IL11RA protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
TPCPQAWGPP GVQYGQPGRP VMLCCPGVNA GTPVSWFRDG DSRLLQGPDS GLGHRLVLAQ VDSRDEGTYV CRTLDGVFGG MVTLKLGSPP ARPEVSCQAV DYENFSCTWS PGRVSGLPTR YLTSYRKKTL PGAESQRESP STGPWPCPQD PLEASRCVVH GAEFWSEYRI NVTEVNPLGA STCLLDVRLQ RILRPDPPQG LRVESVPGYP RRLHASWTYP ASWRRQPHFL LKFRLQYRPA QHPAWSTVEP IGLEELITDA VAGLPHAVRV SARDFLDAGT WSAWSPEAWG TPSTGPLRDE VPDGSRGHEQ KLEAAAQEDS PAPPSPSLQP DPRPLDHRDP LEQVAVLAVE HHHHHH | |
Greater than 85.0% as determined by SDS-PAGE. |