missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL 9 Protein
A cDNA sequence encoding the IL 9 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10721-10ug
Additional Details : Weight : 0.01000kg
Specifications
116558 | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined by a cell proliferation assay using murine TS1 cells is less than 10 ng/ml, corresponding to a specific activity of > 1.0 M 105 IU/mg. | |
Rat | |
Untagged | |
IL9 protein was lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |
IL 9 Protein | |
Research Use Only | |
Il9 | |
Recombinant Protein | |
E. coli | |
MQRCSTSWGI QHTSYLIENL KDDPSSKCSC SANVTSCLCL PIPSDDCTTP CFQEGMSQVT NATQQSKFSP FFFRVKRIVE TLKSNKCQFF SCEKPCNQTT AGNTVSFLKS LLKTFQKTEV QVQRSRA | |
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |