missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL 3 Protein
A cDNA sequence encoding the IL 3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10712-1mg
Additional Details : Weight : 0.01000kg
Specifications
24495 | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MISDRGSDAH HLLRTLDCRT IALEILVKLP YPQVSGLNNS DDKANLRNST LRRVNLDEFL KSQEEFDSQD TTDIKSKLQK LKCCIPAAAS DSVLPGVYNK DLDDFKKKLR FYVIHLKDLQ PVSVSRPPQP TSSSDNFRPM TVEC | |
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
IL 3 Protein | |
Research Use Only | |
Il3 | |
Rat | |
Untagged | |
Interleukin-3 Rat was lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |