missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat ICOS / AILIM / CD278 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) ICOS / AILIM / CD278, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8229-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_072132.1 | |
ICOS / AILIM / CD278 Protein | |
Research Use Only | |
ICOS | |
Recombinant Protein | |
E. coli | |
ELNDLANHRM FSFHDGGVQI SCNYPETVQQ LKMQLFKDRE VLCDLTKTKG SGNTVSIKNP MSCPYQLSNN SVSFFLDNAD SSQGSYFLCS LSIFDPPPFQ EKNLSGGYLL IYESQLCCQL KLWLP | |
Greater than 90% as determined by SDS-PAGE. |
64545 | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |