missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat GMF B Protein
A cDNA sequence encoding the GMF B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12002-10ug
Additional Details : Weight : 0.01000kg
Specifications
GMF B Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
SESLVVCDVA EDLVEKLRKF RFRKETHNAA IIMKIDKDKR LVVLDEELEG VSPDELKDEL PERQPRFIVY SYKYQHDDGR VSYPLCFIFS SPLGCKPEQQ MMYAGSKNKL VQTAELTKVF EIRNTEDLTE EWLREKLGFF H | |
Greater than 97.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Rat | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated (1.0 mg/ml) solution in 1M PBS, pH 7.4. |