missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat GCSF Protein

A cDNA sequence encoding the GCSF was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10625-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15964448

  • £151.37 / 10µg
Estimated Shipment: 10-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

GCSF Protein
10 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
KKIPLLTVSS LPPSLPLPRS FLLKSLEQVR KIQARNTELL EQLCATYKLC HPEELVLFGH SLGIPKASLS SCSSQALQQT KCLSQLHSGL FLYQGLLQAL AGISSELAPT LDMLHLDVDN FATTIWQQME SLGVAPTVQP TQSTMPIFTS AFQRRAGGVL VTSYLQSFLE TAHHALHHLP RPAQKHFPES LFISI
Greater than 97.0% as determined by:(a)Analysis by RP-HPLC. (b)Analysis by SDS-PAGE.
Research Use Only
The ED50 determined by a cell proliferation assay using murine NFS-60 cells is less than 0.05ng/ml, corresponding to a specific activity of > 2.0M 107 IU/mg.
Rat
Untagged
Lyophilized from a 0.2microm filtered solution in 5mM Sodium Citrate, pH 4.0.
Videos
SDS
Documents
Certificates
Special Offers

Special Offers