missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat DPP4 / CD26 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) DPP4 / CD26, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5944-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15948849

  • £2480.00 / 1mg
Estimated Shipment: 10-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_036921.1
DPP4 / CD26 Protein
Research Use Only
Dpp4
Recombinant Protein
E. coli
SMVLGSGSGV FKCGIAVAPV SRWEYYDSVY TERYMGLPTP EDNLDHYRNS TVMSRAENFK QVEYLLIHGT ADDNVHFQQS AQISKALVDA GVDFQAMWYT DEDHGIASST AHQHIYSHMS HFLQQCFSLR
Greater than 90% as determined by SDS-PAGE.
25253
1 mg
Not determined.
Partial (see sequence information for more details).
Rat
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers