missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Cyclophilin A Protein
A cDNA sequence encoding the Cyclophilin A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11578-1mg
Additional Details : Weight : 0.01000kg
Specifications
100360977; 25518 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MKHHHHHHASVNPTVFFDIT ADGEPLGRVC FELFADKVPK TAENFRALST GEKGFGYKGS SFHRIIPGFM CQGGDFTRHN GTGGKSIYGE KFEDENFILK HTGPGILSMA NAGPNTNGSQ FFICTAKTEW LDGKHVVFGK VKEGMSIVEA MERFGSRNGK TSKKITISDC GQL | |
Greater than 95.0% as determined by SDS-PAGE. |
Cyclophilin A Protein | |
Research Use Only | |
Ppia | |
Rat | |
His | |
Cyclophilin-A was filtered (0.4microm) and lyophilized from 0.5 mg/ml solution in phosphate buffered saline and 5%(w/v) trehalose. |