missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat CD74 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) CD74, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5796-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15947829

  • £2475.95 / 1mg
Estimated Shipment: 13-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_037201.1
CD74 Protein
1 mg
Not determined.
Extracellular Domain
Rat
His
Tris-based buffer, 50% glycerol
25599
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Cd74
Recombinant Protein
E. coli
QQQGRLDKLT VTSQNLQLEN LRMKLPKSAK PVSPMRMATP LLMRPLSMDN MLQAPVKNVT KYGNMTQDHV MHLLTKSGPV NYPQLKGSFP ENLKHLKNSM NGLDWKVFES WMKQWLLFEM SKNSLEEKQP TQTPPKVLTK CQEEVSHIPD VHPGAFRPKC DENGNYMPLQ CHGSTGYCWC VFPNGTEVPH TKSRGRHNCS EPLDMEDPSS GLGVTKQDMG QMFL
Videos
SDS
Documents
Certificates
Special Offers

Special Offers