missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat CALB1 Protein
A cDNA sequence encoding the CALB1 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2755.00
Specifications
Gene ID (Entrez) | 83839 |
---|---|
Name | CALB1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | Calb1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15926638
|
enQuireBio™
QP11257-1UG |
1 μg |
£99.15
1µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15936638
|
enQuireBio™
QP11257-5UG |
5 μg |
£155.00
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15916638
|
enQuireBio™
QP11257-100UG |
100 μg |
£2755.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
83839 | |
Research Use Only | |
Calb1 | |
Rat | |
Untagged | |
The protein was lyophilized from a concentrated solution (1 mg/ml) containing 50mM NaHCO3. |
CALB1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MAESHLQSSL ITASQFFEIW LHFDADGSGY LEGKELQNLI QELLQARKKA GLELSPEMKT FVDQYGQRDD GKIGIVELAH VLPTEENFLL LFRCQQLKSC EEFMKTWRKY DTDHSGFIET EELKNFLKDL LEKANKTVDD TKLAEYTDLM LKLFDSNNDG KLELTEMARL LPVQENFLLK FQGIKMCGKE FNKAFELYDQ DGNGYIDENE LDALLKDLCE KNKQELDINN ISTYKKNIMA LSDGGKLYRT DLALILSAGD N | |
Greater than 90% as determined by SDS-PAGE. |