missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Amphoterin-induced protein 1
A DNA sequence encoding the Rattus norvegicus (Rat) Amphoterin-induced protein 3, was expressed in the hosts and tags indicated.
£412.00 - £1165.00
Specifications
Accession Number | NP_835280.1 |
---|---|
Gene ID (Entrez) | 316003 |
Name | Amphoterin-induced protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15952888
|
enQuireBio™
QP10134-MA-20UG |
20 μg |
£412.00
20µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15962888
|
enQuireBio™
QP10134-MA-50UG |
50 μg |
£825.00
50µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15942888
|
enQuireBio™
QP10134-MA-100UG |
100 μg |
£1165.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_835280.1 | |
Amphoterin-induced protein 1 | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |
316003 | |
Research Use Only | |
Amigo3 | |
Recombinant Protein | |
Mammal | |
TSDLEGVLPP DPHNCPNKCV CAADVLSCAG RGLQDLPAAL PATAAELDLS HNALKRLHPG WLAPLSRLRA LYLGYNKLDV LGRGVFTNAS GLRILDLSSN LLRRLRTYDL DGLEELEKLL LFNNRLMHLD LDAFQGLSML SHLYLSCNEL SSFSFNHLHG LGLTRLRTLD LSSNWLGHVS VPELAALPTF LKNRLYLHNN PLPCDCSLYH LLRRWHQRGL SALHDFEREY TCLAFKVAES RVRFFEHSRV FKNCSVAAAP GLELPEEELH THVGQSLRLF CNTTVPAARV AWVSPKNELL VAPGSQDGSI AVLADGSLAI GRVQEQHAGL FVCLASGPRL HHNQTLEYNV SVHKPRPEPE AFNT | |
Greater than 90% as determined by SDS-PAGE. |