missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Amphoterin-induced protein 1

A DNA sequence encoding the Rattus norvegicus (Rat) Amphoterin-induced protein 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP10134-ma-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15962888

  • £825.00 / 50µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_835280.1
Amphoterin-induced protein 1
Research Use Only
Amigo3
Recombinant Protein
Mammal
TSDLEGVLPP DPHNCPNKCV CAADVLSCAG RGLQDLPAAL PATAAELDLS HNALKRLHPG WLAPLSRLRA LYLGYNKLDV LGRGVFTNAS GLRILDLSSN LLRRLRTYDL DGLEELEKLL LFNNRLMHLD LDAFQGLSML SHLYLSCNEL SSFSFNHLHG LGLTRLRTLD LSSNWLGHVS VPELAALPTF LKNRLYLHNN PLPCDCSLYH LLRRWHQRGL SALHDFEREY TCLAFKVAES RVRFFEHSRV FKNCSVAAAP GLELPEEELH THVGQSLRLF CNTTVPAARV AWVSPKNELL VAPGSQDGSI AVLADGSLAI GRVQEQHAGL FVCLASGPRL HHNQTLEYNV SVHKPRPEPE AFNT
Greater than 90% as determined by SDS-PAGE.
316003
50 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers