missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Amphoterin-induced protein 1
A DNA sequence encoding the Rattus norvegicus (Rat) Amphoterin-induced protein 3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP10134-ma-20ug
Additional Details : Weight : 0.01000kg
Specifications
NP_835280.1 | |
Amphoterin-induced protein 1 | |
Research Use Only | |
Amigo3 | |
Recombinant Protein | |
Mammal | |
TSDLEGVLPP DPHNCPNKCV CAADVLSCAG RGLQDLPAAL PATAAELDLS HNALKRLHPG WLAPLSRLRA LYLGYNKLDV LGRGVFTNAS GLRILDLSSN LLRRLRTYDL DGLEELEKLL LFNNRLMHLD LDAFQGLSML SHLYLSCNEL SSFSFNHLHG LGLTRLRTLD LSSNWLGHVS VPELAALPTF LKNRLYLHNN PLPCDCSLYH LLRRWHQRGL SALHDFEREY TCLAFKVAES RVRFFEHSRV FKNCSVAAAP GLELPEEELH THVGQSLRLF CNTTVPAARV AWVSPKNELL VAPGSQDGSI AVLADGSLAI GRVQEQHAGL FVCLASGPRL HHNQTLEYNV SVHKPRPEPE AFNT | |
Greater than 90% as determined by SDS-PAGE. |
316003 | |
20 μg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |