missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Activin A / INHBA Protein

Recombinant Protein

Brand:  enQuireBio™ QP5498-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15966259

  • £77.85 / 2µg
Estimated Shipment: 10-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_058824.1
Activin A / INHBA Protein
2 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Shows bioactivity at a concentration of 10 ng/mL in the Cytotoxicity of MPC-11 cells assay. The bioactivity acceptance criteria is set at 1 x 10^5 units/mg.
Rat
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
29200
Greater than 95% as determined by SDS-PAGE.
Research Use Only
INHBA
Recombinant Protein
E. coli
MGLECDGKVN ICCKKQFFVS FKDIGWNDWI IAPSGYHANY CEGECPSHIA GTSGSSLSFH STVINHYRMR GHSPFANLKS CCVPTKLRPM SMLYYDDGQN IIKKDIQNMI VEECGCS
Videos
SDS
Documents
Certificates
Special Offers

Special Offers