missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant RABV GlycoProtein
A DNA sequence encoding the Rabies virus (strain PM) (RABV) Glycoprotein, was expressed in the hosts and tags indicated.
£292.00 - £2480.00
Specifications
Name | RABV GlycoProtein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | G |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16038750
|
enQuireBio™
QP7413-EC-10UG |
10 μg |
£292.00
10µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16028750
|
enQuireBio™
QP7413-EC-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16048750
|
enQuireBio™
QP7413-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16058750
|
enQuireBio™
QP7413-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16068750
|
enQuireBio™
QP7413-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078750
|
enQuireBio™
QP7413-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
RABV GlycoProtein | |
Not determined. | |
Full Length | |
RABV | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
G | |
Recombinant Protein | |
E. coli | |
KFPIYTILDK LGPWSPIDIH HLSCPNNLVV EDEGCTNLSG FSYMELKVGY ILAIKMNGFT CTGVVTEAET YTNFVGYVTT TFKRKHFRPT PDACRAAYNW KMAGDPRYEE SLHNPYPDYR WLRTVKTTKE SLVIISPSVA DLDPYDRSLH SRVFPSGKCS GVAVSSTYCS TNHDYTIWMP ENPRLGMSCD IFTNSRGKRA SKGSETCGFV DERGLYKSLK GACKLKLCGV LGLRLMDGTW VAMQTSNETK WCPPDQLVNL HDFRSDEIEH LVVEELVRKR EECLDALESI MTTKSVSFRR LSHLRKLVPG FGKAYTIFNK TLMEADAHYK SVRTWNEILP SKGCLRVGGR CHPHVNGVFF NGIILGPDGN VLIPEMQSSL LQQHMELLES SVIPLVHPLA DPSTVFKDGD EAEDFVEVHL PDVHNQVSGV DLGLPNWGKY | |
Greater than 90% as determined by SDS-PAGE. |