missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rabbit TNF a Protein
A cDNA sequence encoding the TNF a was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Gene ID (Entrez) | 100009088 |
---|---|
Name | TNF a Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | TNF |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15955458
|
enQuireBio™
QP10882-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945458
|
enQuireBio™
QP10882-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935458
|
enQuireBio™
QP10882-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
100009088 | |
Research Use Only | |
TNF | |
Recombinant Protein | |
E. coli | |
MSTESMIRDV ELAEGPLPKK AGGPQGSKRC LCLSLFSFLL VAGATTLFCL LHFRVIGPQE EEQSPNNLHL VNPVAQMVTL RSASRALSDK PLAHVVANPQ VEGQLQWLSQ RANALLANGM KLTDNQLVVP ADGLYLIYSQ VLFSGQGCRS YVLLTHTVSR FAVSYPNKVN LLSAIKSPCH RETPEEAEPM AWYEPIYLGG VFQLEKGDRL STEVNQPEYL DLAESGQVYF GIIAL | |
Greater than 95% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
TNF a Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The ED50 as determined by the cytolysis of murine L929 cells in the presence of Actinomycin D is less than 0.03ng/ml, corresponding to a Specific Activity of 30,000,000 IU/mg. | |
Rabbit | |
Untagged | |
TNF-alpha Rabbit was lyophilized after extensive dialysis against 20mM PBS, pH 7.4, 300mM NaCl. |