missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rabbit TNF a Protein

A cDNA sequence encoding the TNF a was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10882-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15955458

  • £99.15 / 5µg
Estimated Shipment: 10-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

100009088
5 μg
< 1.0 EU per ug protein as determined by the LAL method.
The ED50 as determined by the cytolysis of murine L929 cells in the presence of Actinomycin D is less than 0.03ng/ml, corresponding to a Specific Activity of 30,000,000 IU/mg.
Rabbit
Untagged
TNF-alpha Rabbit was lyophilized after extensive dialysis against 20mM PBS, pH 7.4, 300mM NaCl.
TNF a Protein
Research Use Only
TNF
Recombinant Protein
E. coli
MSTESMIRDV ELAEGPLPKK AGGPQGSKRC LCLSLFSFLL VAGATTLFCL LHFRVIGPQE EEQSPNNLHL VNPVAQMVTL RSASRALSDK PLAHVVANPQ VEGQLQWLSQ RANALLANGM KLTDNQLVVP ADGLYLIYSQ VLFSGQGCRS YVLLTHTVSR FAVSYPNKVN LLSAIKSPCH RETPEEAEPM AWYEPIYLGG VFQLEKGDRL STEVNQPEYL DLAESGQVYF GIIAL
Greater than 95% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers