missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rabbit Prolactin Protein

A cDNA sequence encoding the Prolactin was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10850-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15965358

  • £155.00 / 50µg
Estimated Shipment: 03-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Prolactin Protein
Research Use Only
Recombinant Protein
E. coli
MDSKWSRRTG SLLLLLVSNL LLCKSTASLP ICPSGAVNCQ VSLRDLFDRA VILSHHIHKL SSEMFNEFDK RYTQGRGFIT KAINSCHTSS LSTPEDKEQA QQIHHEDLLN MVLRVLPSWN DPLYHLVTEV RGMQEAPDAI LSKAIEIEEQ NRRLLEGMEK IVGQVHPGIK ENEIYSVWSG LPSLQMADED ARLFAFYNLL HCLRRDSHKI DNYLKLLKCR IIYDSNC
Greater than 98.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE.
50 μg
< 1.0 EU per ug protein as determined by the LAL method.
Rabbit
Untagged
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers