missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rabbit PRL R Protein
A cDNA sequence encoding the PRL R was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10843-1mg
Additional Details : Weight : 0.01000kg
Specifications
PRL R Protein | |
Research Use Only | |
Activity is determined by the dose-dependant inhibition of Prolactin-stimuled proliferation of Nb2 cells and by high affinity binding of oPLR and other lactogenic hormones. Refs:1) Bignon et al. (1994) JBC 269; 3318-242) Gertler et al. (1996) JBC 271; 24482-91. | |
Rabbit | |
Untagged | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MKENVASMIV FLLLLFLNIR LLKGQSPPGK PFIFKCRSPE KETFTCWWRP GADGGLPTNY TLTYHKEGET ITHECPDYKT GGPNSCYFSK KHTSIWTIYI ITVNATNQMG SSVSDPRYVD VTYIVEPDPP VNLTLEVKHP EDRKPYLWVK WLPPTLVDVR SGWLTLQYEI RLKPEKAAEW ETHFAGQQTQ FKILSLYPGQ KYLVQVRCKP DHGFWSVWSP ESSIQIPNDF TMKDITVWIF VAVLSTIICL IMVWAVALKG YSMVTCIFPP VPGPKIKGFD THLLEKGKSE ELLSAFGCQD FPPTADCEDL LVEFLEVDDS EDQQLMPAHS KEHSGPGMKP TDLDPDNDSG RGSCDSPSLL SEKCEEPQAN PSTFHTPEVI EQPEKPKANV THTWDPQTIS LVGKMPYLSV NGSKSSTWPL LQPGQHNTNS PYHNIADMCK LATSLDKIDK DALQSSKTTE AAGEEKATKQ REVESSHSKA EQDTGWLLPK EKPPFISPKP LDYVEIHKVN KDGALSLLLK QKENGDQTGK AGTPETSKEY AKVSRVMDNN ILVLVQDPGA QNVALFEEST KEAPPSPSQN QAEKDLSSFS TAPSDCRLQQ GGLDYLDPAC FMHSLH | |
Greater than 97.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |