missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rabbit E-Selectin / CD62e / SELE Protein
A DNA sequence encoding the Oryctolagus cuniculus (Rabbit) E-Selectin / CD62e / SELE, was expressed in the hosts and tags indicated.
£321.05 - £2888.42
Specifications
Accession Number | NP_001075781.1 |
---|---|
Gene ID (Entrez) | 100009151 |
Name | E-Selectin / CD62e / SELE Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16008221
|
enQuireBio™
QP9384-YE-100UG |
100 μg |
£800.00
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16048221
|
enQuireBio™
QP9384-YE-500UG |
500 μg |
£1930.00
500µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16018221
|
enQuireBio™
QP9384-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038221
|
enQuireBio™
QP9384-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058221
|
enQuireBio™
QP9384-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16028221
|
enQuireBio™
QP9384-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001075781.1 | |
E-Selectin / CD62e / SELE Protein | |
Not determined. | |
Extracellular Domain | |
Rabbit | |
His | |
Tris-based buffer, 50% glycerol |
100009151 | |
Research Use Only | |
SELE | |
Recombinant Protein | |
Yeast | |
WTYHFSAENM TYDEASAYCQ QNYTHLVAIQ NKEEIDYLNS ILDYSPSYYW IGIRKVNNVW IWVGTHKPLT EGAKNWAPGE PNNKQNNEDC VEIYIKRPKD TGMWNDERCS KKKLALCYTA ACTEASCSGH GECIETINNY SCKCYPGFSG LKCEQVVTCE AQVQPQHGSL NCTHPLGNFS YNSSCSVSCE RGYLPSSTET TWCTSSGEWS APPATCKVVE CDTMGKPANG DVKCSPSQGS APWNTTCTFD CEEGFTLLGA RSLQCTSSGS WDNEKPTCKA VSCDTIHHPQ NGSVSCSNSS EGKFTFRSSC NFTCEENFLL RGPAQVECTA QGQWTQQAPV CEAVKCDPVH TLEDGFVKCT HPHTGEFTYK SSCTFNCREG FELHGSAQLE CTSQGQWAQE LPSCQVVQCP SLAVLGKTNV SCSGEPVFGT VCNFACPEGW TLNGSAALMC GAEGQWSGML PTCEEPIASN VP | |
Greater than 90% as determined by SDS-PAGE. |