missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rabbit E-Selectin / CD62e / SELE Protein

A DNA sequence encoding the Oryctolagus cuniculus (Rabbit) E-Selectin / CD62e / SELE, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9384-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048221

  • £1698.00 / 500µg

Estimated Shipment: 25-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001075781.1
E-Selectin / CD62e / SELE Protein
500 μg
Not determined.
Extracellular Domain
Rabbit
His
Tris-based buffer, 50% glycerol
100009151
Greater than 90% as determined by SDS-PAGE.
Research Use Only
SELE
Recombinant Protein
Yeast
WTYHFSAENM TYDEASAYCQ QNYTHLVAIQ NKEEIDYLNS ILDYSPSYYW IGIRKVNNVW IWVGTHKPLT EGAKNWAPGE PNNKQNNEDC VEIYIKRPKD TGMWNDERCS KKKLALCYTA ACTEASCSGH GECIETINNY SCKCYPGFSG LKCEQVVTCE AQVQPQHGSL NCTHPLGNFS YNSSCSVSCE RGYLPSSTET TWCTSSGEWS APPATCKVVE CDTMGKPANG DVKCSPSQGS APWNTTCTFD CEEGFTLLGA RSLQCTSSGS WDNEKPTCKA VSCDTIHHPQ NGSVSCSNSS EGKFTFRSSC NFTCEENFLL RGPAQVECTA QGQWTQQAPV CEAVKCDPVH TLEDGFVKCT HPHTGEFTYK SSCTFNCREG FELHGSAQLE CTSQGQWAQE LPSCQVVQCP SLAVLGKTNV SCSGEPVFGT VCNFACPEGW TLNGSAALMC GAEGQWSGML PTCEEPIASN VP
Videos
SDS
Documents
Certificates