missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rabbit CXCL9 / MIG / C-X-C motif chemokine 9 Protein
A DNA sequence encoding the Oryctolagus cuniculus (Rabbit) CXCL9 / MIG / C-X-C motif chemokine 9, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5897-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
CXCL9 / MIG / C-X-C motif chemokine 9 Protein | |
Research Use Only | |
CXCL9 | |
Recombinant Protein | |
E. coli | |
SPIMRNGRCS CISSTQGKIH LQSLKDLKQF SPSPSCGKTE IIATKKDGTQ ICLNPDSTEV KELVEKWKKQ SSPKKKQKKG KKQRKVKKSL KKSQRPHQKK TA | |
Greater than 90% as determined by SDS-PAGE. |
100 μg | |
Not determined. | |
Full Length | |
Rabbit | |
His-SUMO | |
Tris-based buffer, 50% glycerol |