missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Porcine IL 2 Protein

A cDNA sequence encoding the IL 2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10699-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15964688

  • £151.37 / 10µg
Estimated Shipment: 10-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

396868
Greater than 96.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
IL2
Recombinant Protein
E. coli
APTSSSTKNT KKQLEPLLLD LQLLLKEVKN YENADLSRML TFKFYMPKQA TELKHLQCLV EELKALEGVL NLGQSKNSDS ANIKESMNNI NVTVLELKGS ETSFKCEYDD ETVTAVEFLN KWITFCQSIY STLT
IL 2 Protein
10 μg
< 1.0 EU per ug protein as determined by the LAL method.
The ED50 as determined by a cell proliferation assay using murine CTLL-2 cells is less than 0.5ng/ml, corresponding to a specific activity of > 2.0 M 106 IU/mg.
Porcine
Untagged
The protein (1 mg/ml) was lyophilized was lyophilized from a 0.2microm filtered concentrated solution in 1M PBS, pH 7.4.
Videos
SDS
Documents
Certificates
Special Offers

Special Offers